Anti-PDP2 Rabbit Recombinant Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A67901-100UG A67901-50UG
77837-725EA 521.4 CAD
77837-725 77837-726
Anti-PDP2 Rabbit Recombinant Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: PDP2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR