Anti-ECH1 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A55379-100UG A55379-50UG
77824-264EA 521.4 CAD
77824-264 77824-265
Anti-ECH1 Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:20 to 1:200, IF: 1:50 to 1:200

Type:
Antigen: ECH1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR