Anti-PDE8B Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A52755-100UL A52755-50UL
77819-559EA 521.4 CAD
77819-559 77819-560
Anti-PDE8B Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: PDE8B
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR