Anti-XPC Rabbit Recombinant Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A38551-100UG A38551-50UG
77804-746EA 521.4 CAD
77804-746 77804-747
Anti-XPC Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: XPC
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR