Anti-PTPN9 Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A32472-100UG A32472-50UG
77796-137EA 521.4 CAD
77796-137 77796-138
Anti-PTPN9 Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: PTPN9
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR