Anti-Ribosome-inactivating protein PD-L1/PD-L2 Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A50113-100UG A50113-50UG
77814-673EA 521.4 CAD
77814-673 77814-674
Anti-Ribosome-inactivating protein PD-L1/PD-L2 Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: Ribosome-inactivating protein PD-L1/PD-L2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Phytolacca dioica
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR