Anti-III Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A50059-100UG A50059-50UG
77814-578EA 521.4 CAD
77814-578 77814-579
Anti-III Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: III
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Enterobacteria phage M13
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR