Anti-GPR22 Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A66373-100UG A66373-50UG
77834-844EA 521.4 CAD
77834-844 77834-845
Anti-GPR22 Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: GPR22
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR