Anti-SLC19A3 Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A67504-100UG A67504-50UG
77836-968EA 521.4 CAD
77836-968 77836-969
Anti-SLC19A3 Rabbit Recombinant Polyclonal Antibody (HRP)
Type:
Antigen: SLC19A3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR