Anti-NS Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A53973-100UL A53973-50UL
77821-610EA 521.4 CAD
77821-610 77821-611
Anti-NS Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: WB: 1:500 to 1:5000

Type:
Antigen: NS
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Influenza A
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR