Anti-nfuA Rabbit Recombinant Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A52810-100UL A52810-50UL
77819-661EA 521.4 CAD
77819-661 77819-662
Anti-nfuA Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: nfuA
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: E. coli
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR