Anti-VAC14 Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A53928-100UG A53928-50UG
77821-522EA 521.4 CAD
77821-522 77821-523
Anti-VAC14 Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: VAC14
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR