Anti-Dio3 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A51063-100UG A51063-50UG
77816-469EA 521.4 CAD
77816-469 77816-470
Anti-Dio3 Rabbit Recombinant Polyclonal Antibody
Antibodies
Type:
Antigen: Dio3
Clonality: Recombinant Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR