Anti-KIFC1 Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A68079-100UG A68079-50UG
77838-043EA 521.4 CAD
77838-043 77838-044
Anti-KIFC1 Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: KIFC1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR