Anti-egsA Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A52344-100UG A52344-50UG
77818-792EA 521.4 CAD
77818-792 77818-793
Anti-egsA Rabbit Recombinant Polyclonal Antibody
Antibodies
Type:
Antigen: egsA
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Geobacillus stearothermophilus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR