Anti-RASA2 Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A55737-100UL A55737-50UL
77824-950EA 521.4 CAD
77824-950 77824-951
Anti-RASA2 Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: RASA2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR