Anti-PTH2R Rabbit Recombinant Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A32414-100UG A32414-50UG
77796-044EA 521.4 CAD
77796-044 77796-045
Anti-PTH2R Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: PTH2R
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR