Anti-LPAR6 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A30538-100UL A30538-50UL
77793-063EA 521.4 CAD
77793-063 77793-064
Anti-LPAR6 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: WB: 1:1000 to 1:5000, IHC: 1:200 to 1:500, IF: 1:50 to 1:200

Type:
Antigen: LPAR6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR