Anti-SMS Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A35230-100UG A35230-50UG
77800-383EA 521.4 CAD
77800-383 77800-384
Anti-SMS Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: SMS
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
New Product
Order Now

SPECIFICATIONS

Antigen symbol SMS
Environmentally Preferable
Antigen name Spermine synthase protein (16-191AA)
Antigen synonyms MRSR|SMS|Snyder Robinson X linked mental retardation syndrome|Spermidine aminopropyltransferase|Spermine synthase|SPMSY|SpS|SPSY_HUMAN|SRS
Clonality Recombinant Polyclonal
Conjugation FITC (Fluorescein Isothiocyanate)
Reactivity Human
Host Rabbit
Gene ID P52788
Isotype IgG
ELISA Yes
Form Liquid
Purification >95%, Protein G purified
Storage buffer 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
Storage temperature Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
Shipping temperature Shipped on blue ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR