Anti-CEACAM3 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A41194-50UL A41194-100UL
77808-730EA 355.5 CAD
77808-730 77808-729
Anti-CEACAM3 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:2000 to 1:10000, IHC: 1:100 to 1:300

Type:
Antigen: CEACAM3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR