Anti-ARHGAP11B Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: AFG Bioscience
A56416-100UG A56416-50UG
77826-237EA 521.4 CAD
77826-237 77826-238
Anti-ARHGAP11B Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Type:
Antigen: ARHGAP11B
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
New Product
Order Now

SPECIFICATIONS

Antigen symbol ARHGAP11B
Antigen name Rho GTPase-activating protein 11B protein (99-231AA)
Environmentally Preferable
Antigen synonyms ARHGAP11B|FAM7B1|Rho GTPase-activating protein 11B|Protein FAM7B1|Rho-type GTPase-activating protein 11B
Clonality Recombinant Polyclonal
Conjugation FITC (Fluorescein Isothiocyanate)
Reactivity Human
Host Rabbit
Gene ID Q3KRB8
Isotype IgG
ELISA Yes
Form Liquid
Purification >95%, Protein G purified
Storage buffer 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
Storage temperature Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
Shipping temperature Shipped on blue ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR