Anti-WDR41 Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A67141-100UL A67141-50UL
77836-288EA 521.4 CAD
77836-288 77836-289
Anti-WDR41 Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: WDR41
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR