Anti-A4GNT Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A57114-100UL A57114-50UL
77827-517EA 521.4 CAD
77827-517 77827-518
Anti-A4GNT Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:2000 to 1:5000, IHC: 1:25 to 1:100

Type:
Antigen: A4GNT
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
New Product
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR