Anti-HA-33 Rabbit Recombinant Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A50913-100UL A50913-50UL
77816-187EA 521.4 CAD
77816-187 77816-188
Anti-HA-33 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Type:
Antigen: HA-33
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Clostridium botulinum
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR