Anti-DNAL1 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A56730-100UL A56730-50UL
77826-824EA 521.4 CAD
77826-824 77826-825
Anti-DNAL1 Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: WB: 1:200 to 1:1000, IHC: 1:20 to 1:200

Type:
Antigen: DNAL1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR