Anti-MRC2 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A57855-100UL A57855-50UL
77828-903EA 521.4 CAD
77828-903 77828-904
Anti-MRC2 Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: IHC: 1:20 to 1:200

Type:
Antigen: MRC2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR