Anti-PIM3 Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A59608-100UG A59608-50UG
77832-213EA 521.4 CAD
77832-213 77832-214
Anti-PIM3 Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: PIM3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR