Anti-PTPN9 Rabbit Recombinant Antibody

Supplier: AFG Bioscience
A32470-50UG A32470-100UG
77796-134EA 355.5 CAD
77796-134 77796-133
Anti-PTPN9 Rabbit Recombinant Antibody
Antibodies
Recommended dilution: IHC: 1:200 to 1:500

Type:
Antigen: PTPN9
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR