Anti-AGO2 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A49778-100UL A49778-50UL
77814-088EA 521.4 CAD
77814-088 77814-089
Anti-AGO2 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:1000 to 1:2000, IHC: 1:15 to 1:50

Type:
Antigen: AGO2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR