Anti-MS4A8 Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A70067-100UG A70067-50UG
77841-799EA 521.4 CAD
77841-799 77841-800
Anti-MS4A8 Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: MS4A8
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR