Anti-omp Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A57163-100UL A57163-50UL
77827-608EA 521.4 CAD
77827-608 77827-609
Anti-omp Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: omp
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rickettsia japonica
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR