Anti-SH3GL3 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A66495-100UL A66495-50UL
77835-069EA 521.4 CAD
77835-069 77835-070
Anti-SH3GL3 Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: IHC: 1:20 to 1:200

Type:
Antigen: SH3GL3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR