Anti-RBBP4 Rabbit Recombinant Antibody (HRP (Horseradish Peroxidase))

Supplier: AFG Bioscience
A53932-100UG A53932-50UG
77821-529EA 521.4 CAD
77821-529 77821-530
Anti-RBBP4 Rabbit Recombinant Antibody (HRP (Horseradish Peroxidase))
Antibodies
Type:
Antigen: RBBP4
Clonality: Recombinant
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR