Anti-KCNS3 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A66279-100UL A66279-50UL
77834-666EA 521.4 CAD
77834-666 77834-667
Anti-KCNS3 Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: WB: 1:1000 to 1:5000, IHC: 1:20 to 1:200

Type:
Antigen: KCNS3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR