Anti-VN1R1 Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A66680-100UG A66680-50UG
77835-421EA 521.4 CAD
77835-421 77835-422
Anti-VN1R1 Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: VN1R1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR