Anti-GJB7 Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A58902-100UG A58902-50UG
77830-892EA 521.4 CAD
77830-892 77830-893
Anti-GJB7 Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: GJB7
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR