Anti-tar Rabbit Recombinant Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A51370-100UG A51370-50UG
77817-051EA 521.4 CAD
77817-051 77817-052
Anti-tar Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: tar
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Escherichia coli
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR