Anti-USP17L6P Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A58925-100UL A58925-50UL
77830-935EA 521.4 CAD
77830-935 77830-936
Anti-USP17L6P Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: USP17L6P
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR