Anti-CARD16 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A56713-100UL A56713-50UL
77826-790EA 521.4 CAD
77826-790 77826-791
Anti-CARD16 Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: IF: 1:50 to 1:200

Type:
Antigen: CARD16
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR