Anti-MAGI1 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A66093-100UG A66093-50UG
77834-320EA 521.4 CAD
77834-320 77834-321
Anti-MAGI1 Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: IF: 1:50 to 1:200

Type:
Antigen: MAGI1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR