Anti-Alpha-insect toxin LqhaIT Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A50502-100UG A50502-50UG
77815-412EA 521.4 CAD
77815-412 77815-413
Anti-Alpha-insect toxin LqhaIT Rabbit Recombinant Polyclonal Antibody
Antibodies
Type:
Antigen: Alpha-insect toxin LqhaIT
Clonality: Recombinant Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Leiurus quinquestriatus hebraeus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR