Anti-LYPD2 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: AFG Bioscience
A59068-100UG A59068-50UG
77831-201EA 521.4 CAD
77831-201 77831-202
Anti-LYPD2 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Type:
Antigen: LYPD2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR