Anti-AIP Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A52507-100UG A52507-50UG
77819-083EA 521.4 CAD
77819-083 77819-084
Anti-AIP Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: AIP
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR