Anti-ATP2B3 Rabbit Recombinant Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A54309-100UL A54309-50UL
77822-250EA 521.4 CAD
77822-250 77822-251
Anti-ATP2B3 Rabbit Recombinant Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: ATP2B3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR