Anti-ZNF224 Rabbit Recombinant Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A38805-100UG A38805-50UG
77805-163EA 521.4 CAD
77805-163 77805-164
Anti-ZNF224 Rabbit Recombinant Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: ZNF224
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR