Anti-SLC35B1 Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A35012-50UG A35012-100UG
77800-034EA 355.5 CAD
77800-034 77800-033
Anti-SLC35B1 Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: SLC35B1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR