Anti-Ly6e Rabbit Recombinant Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A57410-100UG A57410-50UG
77828-072EA 521.4 CAD
77828-072 77828-073
Anti-Ly6e Rabbit Recombinant Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: Ly6e
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR