Anti-Smox Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A35210-100UG A35210-50UG
77800-347EA 521.4 CAD
77800-347 77800-348
Anti-Smox Rabbit Recombinant Polyclonal Antibody
Antibodies
Type:
Antigen: Smox
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR