Anti-MBD1 Rabbit Recombinant Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A69962-50UG A69962-100UG
77841-598EA 355.5 CAD
77841-598 77841-597
Anti-MBD1 Rabbit Recombinant Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: MBD1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR