Anti-TP73 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: AFG Bioscience
A37268-100UL A37268-50UL
77811-352EA 521.4 CAD
77811-352 77803-352
Anti-TP73 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Type:
Antigen: TP73
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR